Gene Transcripts

Gene Symbol
POGLUT1
Entrez Gene ID
56983
Gene Name
protein O-glucosyltransferase 1
Gene Aliases
C3orf9, CLP46, KDELCL1, KTELC1, MDS010, MDSRP, Rumi, hCLP46
Chromosome Location
Chr. 3: 119468938 - 119494708 on Build GRCh38
UniGene ID
Hs.231750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152305.2 NP_689518.1 2 214
NR_024265.1 2 214
XM_006713705.3 XP_006713768.1 1 25
XM_017006878.1 XP_016862367.1 1 45
XM_017006879.1 XP_016862368.1 2 160
XM_017006880.1 XP_016862369.1 2 160
GenBank mRNA AF168711.1 - 2 165
AK075444.1 - 2 150
AK222937.1 - 2 174
AK295984.1 - 2 189
AK301002.1 - 2 189
AK315367.1 - 2 189
AY298903.1 - 2 200
AY358581.1 - 2 157
BC030614.2 - 2 200 4796951
BC048810.1 - 2 158 5243487
BP347179.1 - 2 214
CD517010.1 - 2 130

View Full Product Details