Gene Transcripts

Gene Symbol
Pdzd8
Entrez Gene ID
107368
Gene Name
PDZ domain containing 8
Gene Aliases
A630041P07Rik, AV329468, AW228944, Pdzk8
Chromosome Location
Chr. 19: 59296084 - 59345780 on Build GRCm38
UniGene ID
Mm.268797
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033222.3 NP_001028394.2 5 1662
GenBank mRNA AK041847.1 - 5 1116
AK145020.1 - 5 1662
BC141370.1 - 5 1567 9055983
BC141371.1 - 5 1567 9055984

View Full Product Details