Gene Transcripts

Gene Symbol
Ifitm6
Entrez Gene ID
213002
Gene Name
interferon induced transmembrane protein 6
Gene Aliases
A330075D07, Dspa2f, Fgls5, fragilis5
Chromosome Location
Chr. 7: 141015812 - 141016892 on Build GRCm38
UniGene ID
Mm.276440
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033632.1 NP_001028804.1 2 526
GenBank mRNA AK039626.1 - 2 542
AY247203.1 - 1 256
BC104338.1 - 2 526 40049250
BC104339.1 - 2 526 40049257

View Full Product Details