Gene Transcripts

Gene Symbol
Slc9a2
Entrez Gene ID
226999
Gene Name
solute carrier family 9 (sodium/hydrogen exchanger), member 2
Gene Aliases
2210416H12Rik, 4932415O19Rik, AV333884, NHE2
Chromosome Location
Chr. 1: 40681712 - 40768885 on Build GRCm38
UniGene ID
Mm.256414
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033289.2 NP_001028461.2 12 2708
GenBank mRNA AK077026.1 - 12 2800
AK138779.1 - 12 3086
BC103685.1 - 12 2708 40048593
BC104737.1 - 12 2708 40048591
BC105676.1 - 12 2707

View Full Product Details