Gene Transcripts

Gene Symbol
Atxn7l1
Entrez Gene ID
380753
Gene Name
ataxin 7-like 1
Gene Aliases
2810423G08Rik, Atxn7l4
Chromosome Location
Chr. 12: 33147639 - 33376408 on Build GRCm38
UniGene ID
Mm.352037
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033436.3 NP_001028608.3 2 325
GenBank mRNA AK036472.1 - 2 275
AK156016.1 - 2 289
AK165286.1 - 2 315
BC100598.1 - 3 427
BC147479.1 - 2 240 9056221
BC147483.1 - 2 240 9056229

View Full Product Details