Gene Transcripts

Gene Symbol
Lrrc8b
Entrez Gene ID
433926
Gene Name
leucine rich repeat containing 8 family, member B
Gene Aliases
R75581, Ta-lrrp, mKIAA0231
Chromosome Location
Chr. 5: 105415775 - 105490071 on Build GRCm38
UniGene ID
Mm.439781
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033550.2 NP_001028722.1 5 2789
GenBank mRNA AK083378.1 - 5 2790

View Full Product Details