Gene Transcripts

Gene Symbol
Mybphl
Entrez Gene ID
68753
Gene Name
myosin binding protein H-like
Gene Aliases
1110037P11Rik
Chromosome Location
Chr. 3: 108364911 - 108380057 on Build GRCm38
UniGene ID
Mm.75201
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026831.1 NP_081107.1 7, 8 1151
GenBank mRNA AK004148.1 - 7, 8 1152
BC089625.1 - 7, 8 1139 6756382
BC132636.1 - 7, 8 1140 40130913
BC132638.1 - 7, 8 1140 40130915

View Full Product Details