Gene Transcripts

Gene Symbol
DERL3
Entrez Gene ID
91319
Gene Name
derlin 3
Gene Aliases
C22orf14, IZP6, LLN2, derlin-3
Chromosome Location
Chr. 22: 23834503 - 23839344 on Build GRCh38
UniGene ID
Hs.593679
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002862.2 NP_001002862.1 7 3090
NM_001135751.1 NP_001129223.1 7 1060
NM_198440.3 NP_940842.2 6 3191

View Full Product Details