Gene Transcripts

Gene Symbol
SPIN3
Entrez Gene ID
169981
Gene Name
spindlin family member 3
Gene Aliases
SPIN-3, TDRD27, bA445O16.1
Chromosome Location
Chr. X: 56976370 - 56995801 on Build GRCh38
UniGene ID
Hs.522672
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027139.1 5 1080
GenBank mRNA AK123122.1 - 6 1274
BC018073.1 - 5 1024
BC032490.1 - 5 1034

View Full Product Details