Gene Transcripts
- Gene Symbol
- ADGRG6
- Entrez Gene ID
- 57211
- Gene Name
- adhesion G protein-coupled receptor G6
- Gene Aliases
- APG1, DREG, GPR126, LCCS9, PS1TP2, VIGR
- Chromosome Location
- Chr. 6: 142301919 - 142446266 on Build GRCh38
- UniGene ID
- Hs.743302
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001032394.2 |
NP_001027566.1
|
15, 16 |
2723 |
|
NM_001032395.2 |
NP_001027567.1
|
15, 16 |
2723 |
|
NM_020455.5 |
NP_065188.4
|
16, 17 |
2807 |
|
NM_198569.2 |
NP_940971.1
|
16, 17 |
2807 |
|
XM_005267061.3 |
XP_005267118.1
|
16, 17 |
2708 |
|
XM_006715516.2 |
XP_006715579.1
|
16, 17 |
2708 |
|
XM_006715517.2 |
XP_006715580.1
|
16, 17 |
2508 |
|
XM_006715518.2 |
XP_006715581.1
|
15, 16 |
2624 |
|
XM_011535964.1 |
XP_011534266.1
|
16, 17 |
2511 |
|
XM_017011085.1 |
XP_016866574.1
|
16, 17 |
2708 |
|
GenBank mRNA |
AB183546.1 |
- |
16, 17 |
2797 |
|
AB183547.1 |
- |
15, 16 |
2713 |
|
AB183548.1 |
- |
16, 17 |
2797 |
|
AB183549.1 |
- |
15, 16 |
2713 |
|
AK027843.1 |
- |
4, 5 |
461 |
|
AY181244.1 |
- |
|
269 |
|
BC075798.1 |
- |
15, 16 |
2783 |
30333185 |
BX640873.1 |
- |
15, 16 |
2329 |
|
BX640971.1 |
- |
16, 17 |
2733 |
|
BX648315.1 |
- |
15, 16 |
2436 |
|
View Full Product Details