Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007203.4 | NP_009134.1 | 7, 8 | 770 | |
NM_147150.2 | NP_671492.1 | 7, 8 | 770 | ||
GenBank mRNA | AB023137.1 | - | 7, 8 | 770 | |
AJ303079.1 | - | 7, 8 | 758 | ||
AJ312217.1 | - | 8, 9 | 1320 | ||
BC140818.1 | - | 7, 8 | 611 | 9021686 | |
BX649126.1 | - | 5, 6 | 556 |