Gene Transcripts

Gene Symbol
Olr98
Entrez Gene ID
293240
Gene Name
olfactory receptor 98
Gene Aliases
Chromosome Location
Chr. 1: 168519499 - 168520446 on Build Rnor_6.0
UniGene ID
Rn.141890
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000143.1 NP_001000143.1 1 827

View Full Product Details