Gene Transcripts

Gene Symbol
Snx17
Entrez Gene ID
298836
Gene Name
sorting nexin 17
Gene Aliases
Chromosome Location
Chr. 6: 26541137 - 26546650 on Build Rnor_6.0
UniGene ID
Rn.162846
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011981.1 NP_001011981.1 15 1517
GenBank mRNA BC078931.1 - 15 1517

View Full Product Details