Gene Transcripts

Gene Symbol
C3orf14
Entrez Gene ID
57415
Gene Name
chromosome 3 open reading frame 14
Gene Aliases
HT021
Chromosome Location
Chr. 3: 62318973 - 62336213 on Build GRCh38
UniGene ID
Hs.47166
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291941.1 NP_001278870.1 4, 5 392
NM_001291942.1 NP_001278871.1 3, 4 280
NM_020685.4 NP_065736.1 4, 5 499
XM_017006931.1 XP_016862420.1 4, 5 875
GenBank mRNA AF236158.1 - 4, 5 329
AK313915.1 - 4, 5 499
BC017772.2 - 3, 4 309 4307022
BU568674.1 - 234
DA783189.1 - 4, 5 392

View Full Product Details