Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014067.2 | NP_001014089.2 | 12 | 1939 | |
XM_006238702.2 | XP_006238764.1 | 11 | 2169 | ||
XM_006238703.2 | XP_006238765.1 | 12 | 2103 | ||
XM_006238704.2 | XP_006238766.1 | 12 | 2097 | ||
XM_006238705.2 | XP_006238767.1 | 11 | 1992 | ||
XM_006238706.2 | XP_006238768.1 | 7 | 1713 | ||
XM_006238707.2 | XP_006238769.1 | 13 | 2031 | ||
GenBank mRNA | BC087059.1 | - | 10 | 2183 |