Gene Transcripts

Gene Symbol
Cited4
Entrez Gene ID
114491
Gene Name
Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4
Gene Aliases
MRG-2, Mrg2
Chromosome Location
Chr. 5: 139597731 - 139598622 on Build Rnor_6.0
UniGene ID
Rn.40380
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053699.1 NP_446151.1 1 694
GenBank mRNA AF361477.1 - 1 694

View Full Product Details