Gene Transcripts

Gene Symbol
H2-DMb2
Entrez Gene ID
15000
Gene Name
histocompatibility 2, class II, locus Mb2
Gene Aliases
H-2Mb2, H2-Mb2
Chromosome Location
Chr. 17: 34145416 - 34151095 on Build GRCm38
UniGene ID
Mm.195060
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010388.4 NP_034518.1 2 251
GenBank mRNA AK154923.1 - 2 355
BC052864.1 - 3 506
BC141105.1 - 2 317 9055717
BC145385.1 - 2 317 9053943
BC145386.1 - 2 317 9053944
BC171960.1 - 2 317 9054246
KJ650224.1 - 2 251
U35334.1 - 2 251
U35335.1 - 2 251
U35336.1 - 2 251
U35337.1 - 2 251
U35338.1 - 2 251
X90796.1 - 2 251
X90797.1 - 2 251
X90798.1 - 2 251
X90799.1 - 2 251
X90800.1 - 2 251
X90801.1 - 2 251
X90802.1 - 2 251
X90803.1 - 2 251
X90804.1 - 2 251
X90805.1 - 2 251
X90806.1 - 2 251
X90807.1 - 2 251
X90808.1 - 2 251

View Full Product Details