Gene Transcripts

Gene Symbol
Sssca1
Entrez Gene ID
56390
Gene Name
Sjogren's syndrome/scleroderma autoantigen 1 homolog (human)
Gene Aliases
1500016H19Rik, C184L
Chromosome Location
Chr. 19: 5730306 - 5731732 on Build GRCm38
UniGene ID
Mm.455818
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020491.5 NP_065237.3 4 682
GenBank mRNA AB012865.1 - 4 650
AK005265.1 - 4 654
BC021593.1 - 4 652 5102521

View Full Product Details