Gene Transcripts

Gene Symbol
Serpina1f
Entrez Gene ID
68348
Gene Name
serine (or cysteine) peptidase inhibitor, clade A, member 1F
Gene Aliases
0610012A11Rik, AI327419, Serpina2, epserin
Chromosome Location
Chr. 12: 103688044 - 103695529 on Build GRCm38
UniGene ID
Mm.20149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB233454.1 - 3 821
BC119526.1 - 2 649 40056096

View Full Product Details