Gene Transcripts

Gene Symbol
Asb7
Entrez Gene ID
117589
Gene Name
ankyrin repeat and SOCS box-containing 7
Gene Aliases
AI449039, Asb-7, D030055C23Rik
Chromosome Location
Chr. 7: 66644565 - 66689650 on Build GRCm38
UniGene ID
Mm.34785
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080443.2 NP_536691.2 4 756
GenBank mRNA AF398968.1 - 1 41
AK051017.1 - 5 744
BC066824.1 - 3 258 30087227
BC085155.1 - 4 736

View Full Product Details