Gene Transcripts
- Gene Symbol
- Tas2r124
- Entrez Gene ID
- 387351
- Gene Name
- taste receptor, type 2, member 124
- Gene Aliases
- T2R24, Tas2r24, mGR24, mt2r50
- Chromosome Location
- Chr. 6: 132754730 - 132755659 on Build GRCm38
- UniGene ID
- Mm.377912
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207026.1 |
NP_996909.1
|
1 |
533 |
|
View Full Product Details