Gene Transcripts

Gene Symbol
EPB41L5
Entrez Gene ID
57669
Gene Name
erythrocyte membrane protein band 4.1 like 5
Gene Aliases
BE37, YMO1, YRT
Chromosome Location
Chr. 2: 120013028 - 120179121 on Build GRCh38
UniGene ID
Hs.369232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184937.1 NP_001171866.1 18, 19 1798
NM_020909.3 NP_065960.2 18, 19 1798
XM_006712651.1 XP_006712714.1 18, 19 1778
XM_006712652.1 XP_006712715.1 18, 19 2093
XM_006712653.1 XP_006712716.1 18, 19 1798
XM_011511529.2 XP_011509831.1 18, 19 1778
XM_017004567.1 XP_016860056.1 18, 19 1634
GenBank mRNA AB046768.1 - 11, 12 1047
AK023019.1 - 18, 19 1749
AK304831.1 - 1, 2 76
AK307458.1 - 1 19
BC054508.1 - 18, 19 1733 5502663

View Full Product Details