Gene Transcripts

Gene Symbol
CD81
Entrez Gene ID
975
Gene Name
CD81 molecule
Gene Aliases
CVID6, S5.7, TAPA1, TSPAN28
Chromosome Location
Chr. 11: 2376177 - 2397419 on Build GRCh38
UniGene ID
Hs.54457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297649.1 NP_001284578.1 6 628
NM_004356.3 NP_004347.1 6 746
XM_011520492.2 XP_011518794.1 6 980
XM_017018598.1 XP_016874087.1 6 489
GenBank mRNA AB209380.1 - 2 3761
BC002978.1 - 6 553 3510634
BC093047.1 - 6 744 30398483
BG397506.1 - 7 616
BT019507.1 - 6 513
BT019508.1 - 6 513
M33680.1 - 6 751

View Full Product Details