Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020859.3 | NP_065910.3 | 11 | 7075 | |
XM_005263163.4 | XP_005263220.1 | 10 | 11529 | ||
XM_005263164.4 | XP_005263221.1 | 10 | 6087 | ||
XM_005263165.4 | XP_005263222.1 | 10 | 7513 | ||
XM_006714282.3 | XP_006714345.1 | 11 | 6230 | ||
XM_011532157.2 | XP_011530459.2 | 10 | 6917 | ||
XM_011532158.2 | XP_011530460.1 | 12 | 6876 | ||
XM_011532159.1 | XP_011530461.1 | 11 | 7065 | ||
XM_011532160.1 | XP_011530462.1 | 9 | 6077 | ||
XM_011532161.2 | XP_011530463.1 | 10 | 6255 | ||
XM_017008488.1 | XP_016863977.1 | 11 | 6233 | ||
XM_017008489.1 | XP_016863978.1 | 11 | 6134 | ||
GenBank mRNA | AB040914.1 | - | 7 | 4274 | |
AB055660.1 | - | 11 | 7076 | ||
AF109367.1 | - | 2 | 728 | ||
AK128677.1 | - | 7 | 5577 | ||
BC007291.1 | - | 7 | 2436 |