Gene Transcripts

Gene Symbol
STK35
Entrez Gene ID
140901
Gene Name
serine/threonine kinase 35
Gene Aliases
CLIK1, STK35L1
Chromosome Location
Chr. 20: 2098035 - 2148555 on Build GRCh38
UniGene ID
Hs.100057
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080836.3 NP_543026.2 4 4258
XM_011529174.2 XP_011527476.1 3 7355
GenBank mRNA AK024394.1 - 1 2476

View Full Product Details