Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_130896.2 | NP_570966.2 | 5, 6 | 671 | |
NM_181510.2 | NP_852611.2 | 5, 6 | 671 | ||
GenBank mRNA | AF492015.1 | - | 5, 6 | 671 | |
AF492016.1 | - | 5, 6 | 671 | ||
AL591715.1 | - | 4, 5 | 566 | ||
GQ472209.1 | - | 5, 6 | 671 |