Gene Transcripts

Gene Symbol
CRIP3
Entrez Gene ID
401262
Gene Name
cysteine rich protein 3
Gene Aliases
TLP, TLP-A, bA480N24.2
Chromosome Location
Chr. 6: 43305473 - 43310705 on Build GRCh38
UniGene ID
Hs.653165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206922.2 NP_996805.2 1, 2 39
XM_005249103.3 XP_005249160.1 1, 2 1952
XM_011514609.2 XP_011512911.1 1, 2 1952
XM_017010856.1 XP_016866345.1 1, 2 1952
XM_017010857.1 XP_016866346.1 1, 2 1952
GenBank mRNA AY555741.1 - 1, 2 39
AY555742.1 - 1, 2 39
AY555743.1 - 1, 2 39
AY555744.1 - 1, 2 39

View Full Product Details