Gene Transcripts

Gene Symbol
NPW
Entrez Gene ID
283869
Gene Name
neuropeptide W
Gene Aliases
L8, L8C, PPL8, PPNPW
Chromosome Location
Chr. 16: 2019520 - 2020755 on Build GRCh38
UniGene ID
Hs.233533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099456.2 NP_001092926.2 1 768
GenBank mRNA AB084276.1 - 1 548
AL365519.1 - 1 88
BC150534.1 - 1 272
BC171886.1 - 1 985

View Full Product Details