Gene Transcripts

Gene Symbol
PAGE2
Entrez Gene ID
203569
Gene Name
PAGE family member 2
Gene Aliases
CT16.4, GAGEC2, GAGEE2, PAGE-2
Chromosome Location
Chr. X: 55089052 - 55092836 on Build GRCh38
UniGene ID
Hs.662489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207339.3 NP_997222.1 3, 4 246
XM_017029353.1 XP_016884842.1 3, 4 275
GenBank mRNA BC054022.1 - 3, 4 240

View Full Product Details