Gene Transcripts

Gene Symbol
GPR78
Entrez Gene ID
27201
Gene Name
G protein-coupled receptor 78
Gene Aliases
Chromosome Location
Chr. 4: 8580237 - 8590023 on Build GRCh38
UniGene ID
Hs.740355
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080819.4 NP_543009.2 2 1220
NR_045511.2 2 527
GenBank mRNA AF411107.1 - 2 727
AK128807.1 - 2 1220
AK307281.1 - 2 527
AK313038.1 - 2 795
AY359107.1 - 2 1021
BC057778.1 - 2 1144 30343110
BC069343.1 - 2 727 7262307
BC069813.1 - 2 727 7262319
JF810885.1 - 2 727

View Full Product Details