Gene Transcripts

Gene Symbol
PARP12
Entrez Gene ID
64761
Gene Name
poly(ADP-ribose) polymerase family member 12
Gene Aliases
ARTD12, MST109, MSTP109, ZC3H1, ZC3HDC1
Chromosome Location
Chr. 7: 140023744 - 140063721 on Build GRCh38
UniGene ID
Hs.12646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022750.3 NP_073587.1 4 1692
NR_130117.1 4 1692
XM_005250038.3 XP_005250095.1 4 1687
XM_005250039.4 XP_005250096.1 4 1689
XM_005250040.4 XP_005250097.1 4 1689
XM_011516491.2 XP_011514793.1 4 1687
GenBank mRNA AL136766.1 - 4 1314
AL137255.1 - 4 511
BC081541.1 - 4 1692 5296560

View Full Product Details