Gene Transcripts
- Gene Symbol
- ATG9A
- Entrez Gene ID
- 79065
- Gene Name
- autophagy related 9A
- Gene Aliases
- APG9L1, MGD3208, mATG9
- Chromosome Location
- Chr. 2: 219219380 - 219229688 on Build GRCh38
- UniGene ID
- Hs.323363
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001077198.2 |
NP_001070666.1
|
9, 10 |
1659 |
|
NM_024085.4 |
NP_076990.4
|
8, 9 |
1607 |
|
GenBank mRNA |
AK025822.1 |
- |
3, 4 |
961 |
|
AK056595.1 |
- |
9, 10 |
1600 |
|
AK289755.1 |
- |
9, 10 |
1554 |
|
AK302521.1 |
- |
8, 9 |
1498 |
|
AK315553.1 |
- |
3, 4 |
971 |
|
AL833865.1 |
- |
2, 3 |
856 |
|
BC001098.2 |
- |
1, 2 |
134 |
|
BX537984.1 |
- |
8, 9 |
1528 |
|
BX538192.1 |
- |
8, 9 |
1502 |
|
BX538198.1 |
- |
9, 10 |
1436 |
|
View Full Product Details