Gene Transcripts
- Gene Symbol
- LILRP2
- Entrez Gene ID
- 79166
- Gene Name
- leukocyte immunoglobulin-like receptor pseudogene 2
- Gene Aliases
- CD85m, ILT10, LILRA5
- Chromosome Location
- Chr. 19: 54707916 - 54713452 on Build GRCh38
- UniGene ID
- Hs.202680
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003061.2 |
|
2 |
629 |
|
GenBank mRNA |
BC131616.1 |
- |
2 |
629 |
|
View Full Product Details