Gene Transcripts

Gene Symbol
LENG1
Entrez Gene ID
79165
Gene Name
leukocyte receptor cluster member 1
Gene Aliases
Chromosome Location
Chr. 19: 54155639 - 54159743 on Build GRCh38
UniGene ID
Hs.590974
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024316.2 NP_077292.2 2 308
GenBank mRNA AF211966.1 - 1 86
BC014986.1 - 2 273 4877885
BP870984.1 - 2 309

View Full Product Details