Gene Transcripts

Gene Symbol
THOC6
Entrez Gene ID
79228
Gene Name
THO complex 6
Gene Aliases
BBIS, WDR58, fSAP35
Chromosome Location
Chr. 16: 3024031 - 3027755 on Build GRCh38
UniGene ID
Hs.412304
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142350.1 NP_001135822.1 6, 7 715
NM_024339.3 NP_077315.2 6, 7 715
GenBank mRNA AK075330.1 - 6, 7 682
AK308960.1 - 3, 4 961
AK314086.1 - 6, 7 693
AL832548.1 - 5, 6 319
BC003118.1 - 7, 8 595 3537243
BC050674.1 - 6, 7 654 6067702

View Full Product Details