Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111322.1 | NP_001104792.1 | 18 | 2271 | |
NM_024072.3 | NP_076977.3 | 18 | 2271 | ||
GenBank mRNA | AF478457.1 | - | 18 | 2271 | |
AY148094.1 | - | 18 | 2250 | ||
BC005848.2 | - | 18 | 2192 |