Gene Transcripts

Gene Symbol
TCTN1
Entrez Gene ID
79600
Gene Name
tectonic family member 1
Gene Aliases
JBTS13, TECT1
Chromosome Location
Chr. 12: 110614027 - 110649130 on Build GRCh38
UniGene ID
Hs.211511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024549.5 NP_078825.2 6, 7 933
XM_011538733.2 XP_011537035.1 6, 7 837
XM_017019969.1 XP_016875458.1 6, 7 802
GenBank mRNA AK055891.1 - 6, 7 847
BC040113.1 - 6, 7 823 6054789

View Full Product Details