Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024766.4 | NP_079042.1 | 10 | 905 | |
XM_011533113.2 | XP_011531415.1 | 8 | 581 | ||
XM_017004982.1 | XP_016860471.1 | 8 | 3008 | ||
XM_017004983.1 | XP_016860472.1 | 9 | 1120 | ||
GenBank mRNA | AK027104.1 | - | 10 | 845 | |
BC053733.1 | - | 10 | 832 | 5199179 |