Gene Transcripts

Gene Symbol
C2orf54
Entrez Gene ID
79919
Gene Name
chromosome 2 open reading frame 54
Gene Aliases
Chromosome Location
Chr. 2: 240886048 - 240896889 on Build GRCh38
UniGene ID
Hs.193745
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK026324.1 - 2 275
BC125154.1 - 2 266 40120634
BC125155.1 - 2 266 40120635

View Full Product Details