Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114133.2 | NP_001107605.1 | 4 | 4256 | |
NM_024875.4 | NP_079151.2 | 2 | 3433 | ||
XM_005270158.2 | XP_005270215.1 | 2 | 3755 | ||
XM_005270159.3 | XP_005270216.1 | 2 | 3389 | ||
GenBank mRNA | AB188489.1 | - | 4 | 4223 | |
AK022983.1 | - | 2 | 3425 | ||
W92641.1 | - | 429 |