Gene Transcripts
- Gene Symbol
- RPS2P45
- Entrez Gene ID
- 80052
- Gene Name
- ribosomal protein S2 pseudogene 45
- Gene Aliases
- RPS2_21_1492
- Chromosome Location
- Chr. 16: 69033630 - 69034561 on Build GRCh38
- UniGene ID
- Hs.661121
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC104819.1 |
- |
1 |
154 |
|
BC104821.1 |
- |
1 |
154 |
|
View Full Product Details