Gene Transcripts
- Gene Symbol
- THOC7
- Entrez Gene ID
- 80145
- Gene Name
- THO complex 7
- Gene Aliases
- NIF3L1BP1, fSAP24, hTREX30
- Chromosome Location
- Chr. 3: 63833870 - 63864478 on Build GRCh38
- UniGene ID
- Hs.288151
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK027098.1 |
- |
2 |
118 |
|
View Full Product Details