Gene Transcripts
- Gene Symbol
- PRR3
- Entrez Gene ID
- 80742
- Gene Name
- proline rich 3
- Gene Aliases
- CAT56
- Chromosome Location
- Chr. 6: 30556709 - 30564696 on Build GRCh38
- UniGene ID
- Hs.118354
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001077497.2 |
NP_001070965.1
|
2, 3 |
1031 |
|
NM_025263.3 |
NP_079539.2
|
3, 4 |
1094 |
|
GenBank mRNA |
AK074531.1 |
- |
2, 3 |
460 |
|
AK131382.1 |
- |
1, 2 |
2422 |
|
AK298621.1 |
- |
3, 4 |
818 |
|
AK316586.1 |
- |
3, 4 |
823 |
|
BC126455.1 |
- |
3, 4 |
674 |
8992171 |
BC126457.1 |
- |
3, 4 |
674 |
8992173 |
CN258740.1 |
- |
3, 4 |
487 |
|
DR762096.1 |
- |
2, 3 |
421 |
|
U63336.1 |
- |
3, 4 |
917 |
|
View Full Product Details