Gene Transcripts

Gene Symbol
SLC25A21
Entrez Gene ID
89874
Gene Name
solute carrier family 25 member 21
Gene Aliases
ODC, ODC1
Chromosome Location
Chr. 14: 36677921 - 37172660 on Build GRCh38
UniGene ID
Hs.730857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171170.1 NP_001164641.1 1, 2 388
NM_030631.3 NP_085134.1 1, 2 388
GenBank mRNA AJ278148.1 - 1, 2 388
AK289575.1 - 1, 2 151
BC101521.1 - 1, 2 124 8069027
BC113365.1 - 1, 2 124 8322417

View Full Product Details