Gene Transcripts

Gene Symbol
RSPH6A
Entrez Gene ID
81492
Gene Name
radial spoke head 6 homolog A
Gene Aliases
RSHL1, RSP4, RSP6, RSPH4B
Chromosome Location
Chr. 19: 45795710 - 45815347 on Build GRCh38
UniGene ID
Hs.146544
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030785.3 NP_110412.1 6 2457
GenBank mRNA AK093068.1 - 5 1727
AK098837.1 - 6 2402
AL136761.1 - 6 2429
BC057785.1 - 6 2285

View Full Product Details