Gene Transcripts
- Gene Symbol
- PITPNM3
- Entrez Gene ID
- 83394
- Gene Name
- PITPNM family member 3
- Gene Aliases
- ACKR6, CORD5, NIR1, RDGBA3
- Chromosome Location
- Chr. 17: 6451263 - 6556557 on Build GRCh38
- UniGene ID
- Hs.183983
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001165966.1 |
NP_001159438.1
|
14, 15 |
2058 |
|
NM_031220.3 |
NP_112497.2
|
15, 16 |
2166 |
|
XM_011524015.2 |
XP_011522317.1
|
15, 16 |
2106 |
|
XM_011524016.2 |
XP_011522318.1
|
15, 16 |
2107 |
|
GenBank mRNA |
AB209130.1 |
- |
9, 10 |
4529 |
|
AF334586.1 |
- |
15, 16 |
2163 |
|
AL389994.1 |
- |
14, 15 |
1889 |
|
BC020484.1 |
- |
6, 7 |
764 |
|
BC128583.1 |
- |
15, 16 |
2103 |
40128716 |
BC128584.1 |
- |
15, 16 |
2103 |
40128724 |
View Full Product Details