Gene Transcripts

Gene Symbol
AMMECR1L
Entrez Gene ID
83607
Gene Name
AMMECR1 like
Gene Aliases
Chromosome Location
Chr. 2: 127861630 - 127886408 on Build GRCh38
UniGene ID
Hs.546449
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199140.1 NP_001186069.1 8 3826
NM_031445.2 NP_113633.2 8 3798
XM_005263804.4 XP_005263861.1 9 4429
XM_005263806.4 XP_005263863.1 8 4318
XM_005263807.4 XP_005263864.1 7 4183
XM_011511952.2 XP_011510254.1 8 4319
XM_011511954.2 XP_011510256.1 9 4406
XM_011511955.2 XP_011510257.1 7 3348
GenBank mRNA AK024993.1 - 1 1273
AK026685.1 - 1 2159
BC016181.1 - 8 3798 4644159
BC078174.1 - 8 3808 6012672
BC084552.1 - 1 146

View Full Product Details