Gene Transcripts

Gene Symbol
GORASP1
Entrez Gene ID
64689
Gene Name
golgi reassembly stacking protein 1
Gene Aliases
GOLPH5, GRASP65, P65
Chromosome Location
Chr. 3: 39096599 - 39108363 on Build GRCh38
UniGene ID
Hs.721946
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031899.3 NP_114105.1 3 980
NR_103866.1 3 517
NR_103867.1 3 517
XM_006713301.3 XP_006713364.1 3 1080
XM_011534020.2 XP_011532322.1 3 1080
XM_011534021.2 XP_011532323.1 3 1080
XM_017007050.1 XP_016862539.1 3 256
XM_017007051.1 XP_016862540.1 3 695
XM_017007052.1 XP_016862541.1 3 242
GenBank mRNA AJ409349.1 - 3 256
AK001574.1 - 3 255
AK091168.1 - 3 444
AK124755.1 - 3 980
AK300271.1 - 3 196
BC008928.2 - 3 238
BC075854.1 - 3 266 5503263
BC103826.1 - 3 198 40001584
BC103827.1 - 3 198 40001588
DB031221.1 - 3 255

View Full Product Details