Gene Transcripts

Gene Symbol
IRF9
Entrez Gene ID
10379
Gene Name
interferon regulatory factor 9
Gene Aliases
IRF-9, ISGF3, ISGF3G, p48
Chromosome Location
Chr. 14: 24161213 - 24166565 on Build GRCh38
UniGene ID
Hs.1706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006084.4 NP_006075.3 9 1547
GenBank mRNA BC035716.2 - 8 1423
BI497082.1 - 1 287
M87503.1 - 9 1455

View Full Product Details