Gene Transcripts
- Gene Symbol
- WDR54
- Entrez Gene ID
- 84058
- Gene Name
- WD repeat domain 54
- Gene Aliases
- Chromosome Location
- Chr. 2: 74421721 - 74425755 on Build GRCh38
- UniGene ID
- Hs.643480
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320823.1 |
NP_001307752.1
|
3, 4 |
358 |
|
NM_001320824.1 |
NP_001307753.1
|
3, 4 |
413 |
|
NM_001320825.1 |
NP_001307754.1
|
2, 3 |
157 |
|
NM_032118.3 |
NP_115494.1
|
3, 4 |
380 |
|
XM_017005064.1 |
XP_016860553.1
|
2, 3 |
159 |
|
GenBank mRNA |
AK023015.1 |
- |
3, 4 |
373 |
|
AK222696.1 |
- |
3, 4 |
373 |
|
BC051848.1 |
- |
3, 4 |
345 |
6150670 |
BG024606.1 |
- |
3, 4 |
379 |
|
BI855898.1 |
- |
3, 4 |
321 |
|
CD249537.1 |
- |
2, 3 |
173 |
|
HY088750.1 |
- |
3, 4 |
395 |
|
View Full Product Details